1,3-Dimethoxybenzene 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Resorcinol dimethyl ether

OR1557-100G OR1557-500G
77405-466EA 21.32 USD
77405-466 77405-470
1,3-Dimethoxybenzene 98%
1,3-Dimethoxybenzene
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 217 °C
Melting Pt: –52 °C
Density: 1.05 g/cm³ (25 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00008384
CAS Number: 151-10-0
EINECS: 205-783-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR