4-(Chloromethyl)thiazole hydrochloride
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR15568-100G
OR15568-25G
OR15568-5G
77405-454EA
461.52
USD
77405-454
77405-456
77405-458
4-(Chloromethyl)thiazole hydrochloride
4-(Chloromethyl)thiazole hydrochloride
Formula:
C₄H₄ClNS Storage Temperature: Ambient |
MDL Number:
MFCD02094738 CAS Number: 7709-58-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...