5-Bromo-2-ethoxypyrimidine

Supplier: APOLLO SCIENTIFIC
Warning

OR15514-1G OR15514-5G
77405-428EA 116.14 USD
77405-428 77405-430
5-Bromo-2-ethoxypyrimidine
5-Bromo-2-ethoxypyrimidine
Formula: C₆H₇BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD07375135
CAS Number: 17758-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR