4-(Ethylsulfonyl)benzaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

OR2727-1G OR2727-250MG
77407-922EA 131.42 USD
77407-922 77407-924
4-(Ethylsulfonyl)benzaldehyde
4-(Ethylsulfonyl)benzaldehyde
Formula: C₉H₁₀O₃S
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD09264545
CAS Number: 50899-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR