8-Hydroxyjulolidine 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1,2,3,5,6,7-Hexahydropyrido[3,2,1-ij]quinolin-8-ol, 2,3,6,7-Tetrahydro-1H,5H-benzo[ij]quinolizin-8-ol

OR24664-1G OR24664-5G
77407-522EA 103.92 USD
77407-522 77407-524
8-Hydroxyjulolidine 97%
8-Hydroxyjulolidine
Formula: C₁₂H₁₅NO
MW: 189.26 g/mol
Melting Pt: 132…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00006918
CAS Number: 41175-50-2
EINECS: 255-247-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR