1-Phenylimidazole 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR24638-1G OR24638-5G OR24638-10G
77407-514EA 21.4 USD
77407-514 77407-516 77407-512
1-Phenylimidazole 98%
1-Phenylimidazole
Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 142 °C/15 mmHg
Melting Pt: 13 °C
Density: 1.14 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00041204
CAS Number: 7164-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR