2-Bromopyrimidine

Supplier: APOLLO SCIENTIFIC
Warning

OR23100-100G OR23100-25G OR23100-5G
77407-104EA 141.62 USD
77407-104 77407-106 77407-108
2-Bromopyrimidine
2-Bromopyrimidine
Formula: C₄H₃BrN₂
Boiling Pt: 62…64 °C/1.5 mmHg
Melting Pt: 53…57 °C
Storage Temperature: Freezer
MDL Number: MFCD00014601
CAS Number: 4595-60-2
EINECS: 224-993-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR