Benzo[b]furan-2-boronic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Benzofuran-2-boronic acid, 2-Benzofurylboronic acid

OR23097-100G OR23097-25G OR23097-5G
77407-094EA 447.26 USD
77407-094 77407-096 77407-098
Benzo[b]furan-2-boronic acid 98%
Benzo[b]furan-2-boronic acid
Formula: C₈H₇BO₃
MW: 161.95 g/mol
Melting Pt: 196…198 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00236019
CAS Number: 98437-24-2
New Product

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR