3-Furoic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Furan-3-carboxylic acid, 3-Furancarboxylic acid

OR23027-100G OR23027-25G OR23027-5G
77407-014EA 165.04 USD
77407-014 77407-016 77407-018
3-Furoic acid 98%
3-Furoic acid
Formula: C₅H₄O₃
MW: 112.08 g/mol
Melting Pt: 120…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00005350
CAS Number: 488-93-7
EINECS: 207-689-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR