3-Bromo-2-thiophenecarbonitrile 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Bromothiophene-2-carbonitrile

OR18012-1G OR18012-250MG OR18012-5G
77406-176EA 36.68 USD
77406-176 77406-178 77406-180
3-Bromo-2-thiophenecarbonitrile 95%
3-Bromo-2-thiophenecarbonitrile
Formula: C₅H₂BrNS
MW: 188.05 g/mol
Melting Pt: 43…48 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00068173
CAS Number: 18791-98-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR