tert-Butyl-2,8-diazaspiro[4.5]decane-2-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: tert-Butyl 2,8-diazaspiro[4.5]decane-2-carboxylate, 2,8-Diazaspiro[4.5]decane, N2-BOC protected

OR16902-250MG OR16902-500MG
77405-842EA 73.36 USD
77405-842 77405-844
tert-Butyl-2,8-diazaspiro[4.5]decane-2-carboxylate
tert-Butyl-2,8-diazaspiro[4.5]decane-2-carboxylate
Formula: C₁₃H₂₄N₂O₂
MW: 240.3419 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06739056
CAS Number: 336191-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR