4-Methoxybenzyl bromide, Technical Grade

Supplier: APOLLO SCIENTIFIC
Danger

OR16607-1G OR16607-25G OR16607-5G
77405-760EA 50.94 USD
77405-760 77405-762 77405-764
4-Methoxybenzyl bromide, Technical Grade
4-Methoxybenzyl bromide
Formula: C₈H₉BrO
Boiling Pt: 91 °C (1 mmHg)
Density: 1.395 g/cm³ (25 °C)
Storage Temperature: Freezer
MDL Number: MFCD00800380
CAS Number: 2746-25-0
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR