1,10-Phenanthroline-5,6-dione 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR16008-1G OR16008-25G OR16008-5G
77405-604EA 55.02 USD
77405-604 77405-606 77405-608
1,10-Phenanthroline-5,6-dione 98%
1,10-Phenanthroline-5,6-dione
Formula: C₁₂H₆N₂O₂
Melting Pt: 260 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00014473
CAS Number: 27318-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR