5-(Pyrrolidin-1-yl)pyridine-2-carboxylic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR15955-1G OR15955-250MG
77405-594EA 410.58 USD
77405-594 77405-596
5-(Pyrrolidin-1-yl)pyridine-2-carboxylic acid 95%
5-(Pyrrolidin-1-yl)pyridine-2-carboxylic acid
Formula: C₁₀H₁₂N₂O₂
Melting Pt: 193.5…197 °C
Storage Temperature: Ambient
MDL Number: MFCD09879903
CAS Number: 950603-19-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR