5,5-Dimethyl-1-pyrroline-N-oxide 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR16990-10G
OR16990-1G
OR16990-5G
77405-886EA
256.74
USD
77405-886
77405-888
77405-890
5,5-Dimethyl-1-pyrroline-N-oxide 97%
5,5-Dimethyl-1-pyrroline-N-oxide
Formula:
C₆H₁₁NO Boiling Pt: 78 °C/1.4 mmHg Melting Pt: 25…29 °C Density: 1.015 g/cm³ (25 °C) Flash Pt: 95 °C Storage Temperature: Freezer |
MDL Number:
MFCD00005279 CAS Number: 3317-61-1 EINECS: 222-011-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...