Hydroxyterephthalic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR16986-1G OR16986-25G OR16986-5G
77405-874EA 49.92 USD
77405-874 77405-876 77405-878
Hydroxyterephthalic acid 97%
Hydroxyterephthalic acid
Formula: C₈H₆O₅
Melting Pt: 312…317 °C
Storage Temperature: Ambient
MDL Number: MFCD09835368
CAS Number: 636-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR