4-Amino-2-methoxypyrimidine 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR16620-1G OR16620-5G
77405-776EA 28.52 USD
77405-776 77405-778
4-Amino-2-methoxypyrimidine 97%
4-Amino-2-methoxypyrimidine
Formula: C₅H₇N₃O
Storage Temperature: Ambient
MDL Number: MFCD00128201
CAS Number: 3289-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR