N-Methyl-1-(p-tolyl)methanamine

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: N-Methyl-4-methylbenzylamine

OR16470-1G OR16470-25G OR16470-5G
77405-704EA 55.02 USD
77405-704 77405-706 77405-708
N-Methyl-1-(p-tolyl)methanamine
N-Methyl-1-(p-tolyl)methanamine
Formula: C₉H₁₃N
MW: 135.2100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115412
CAS Number: 699-04-7
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR