2-Hydroxy-5-iodonicotinic acid 95%

Supplier: APOLLO SCIENTIFIC

OR16444-5G OR16444-1G
77405-694EA 199.7 USD
77405-694 77405-692
2-Hydroxy-5-iodonicotinic acid 95%
2-Hydroxy-5-iodonicotinic acid
Formula: C₆H₅NO₃
MW: 265.005 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11100199
CAS Number: 390360-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR