2-Butyl-1,3-benzoxazol-4-amine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Amino-2-(but-1-yl)-1,3-benzoxazole

OR15326-100MG OR15326-250MG
77405-364EA 50.94 USD
77405-364 77405-366
2-Butyl-1,3-benzoxazol-4-amine
2-Butyl-1,3-benzoxazol-4-amine
Formula: C₁₁H₁₄N₂O
Storage Temperature: Ambient
CAS Number: 886361-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR