Ethyl-2-(3,4-dimethoxyphenyl)-4-hydroxy-1,3-thiazole-5-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

OR15091-100MG OR15091-500MG
77405-282EA 54.26 USD
77405-282 77405-284
Ethyl-2-(3,4-dimethoxyphenyl)-4-hydroxy-1,3-thiazole-5-carboxylate
Ethyl-2-(3,4-dimethoxyphenyl)-4-hydroxy-1,3-thiazole-5-carboxylate
Formula: C₁₄H₁₅NO₅S
MW: 309.34 g/mol
Melting Pt: 144…146 °C
Storage Temperature: Ambient
MDL Number: MFCD08444004
CAS Number: 924868-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR