4-(3,5-Dimethyl-1H-pyrazol-1-yl)-N-methylbenzylamine 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-[4-(3,5-Dimethyl-1H-pyrazol-1-yl)phenyl]-N-methylmethylamine, 3,5-Dimethyl-1-{4-[(methylamino)methyl]phenyl}-1H-pyrazole

OR14913-1G OR14913-250MG
77405-174EA 294.44 USD
77405-174 77405-176
4-(3,5-Dimethyl-1H-pyrazol-1-yl)-N-methylbenzylamine 95%
4-(3,5-Dimethyl-1H-pyrazol-1-yl)-N-methylbenzylamine
Formula: C₁₃H₁₇N₃
MW: 215.294 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08572134
CAS Number: 937796-07-1
UN: 2922
ADR: 8+6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR