4-Methyl-1H-1,2,3-benzotriazole 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Methyl-1H-benzo[d][1,2,3]triazole

OR18853-1G OR18853-5G
77406-418EA 316.86 USD
77406-418 77406-420
4-Methyl-1H-1,2,3-benzotriazole 95%
4-Methyl-1H-1,2,3-benzotriazole
Formula: C₇H₇N₃
MW: 133.15 g/mol
Melting Pt: 141…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00035800
CAS Number: 29878-31-7
EINECS: 249-921-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR