3-Iodo-4-methyl-1H-indazole

Supplier: APOLLO SCIENTIFIC
Warning

OR18424-5G OR18424-1G
77406-334EA 191.54 USD
77406-334 77406-332
3-Iodo-4-methyl-1H-indazole
3-Iodo-4-methyl-1H-indazole
Formula: C₈H₇IN₂
MW: 258.0591 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781537
CAS Number: 885522-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR