5-Bromothiophene-2-carbaldehyde 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR18347-100G OR18347-1G OR18347-25G
77406-264EA 136.52 USD
77406-264 77406-266 77406-268
5-Bromothiophene-2-carbaldehyde 97%
5-Bromothiophene-2-carbaldehyde
Formula: C₅H₃BrOS
Boiling Pt: 105…107 °C/11 mmHg
Melting Pt: 15 °C
Density: 1.607 g/cm³ (25 °C)
Flash Pt: 99 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005432
CAS Number: 4701-17-1
EINECS: 225-176-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR