2-Aminobenzothiazole 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Benzothiazol-2-ylamine, Benzo[d]thiazol-2-amine, Cerium(III)-Ionophore II

OR18345-100G OR18345-25G OR18345-500G
77406-254EA 34.64 USD
77406-254 77406-256 77406-258
2-Aminobenzothiazole 97%
2-Aminobenzothiazole
Formula: C₇H₆N₂S
MW: 150.2 g/mol
Melting Pt: 126…129 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005785
CAS Number: 136-95-8
EINECS: 205-268-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR