7-Methoxy-1-tetralone 97%

Supplier: APOLLO SCIENTIFIC

OR17678-100G OR17678-25G OR17678-500G
77406-082EA 116.14 USD
77406-082 77406-084 77406-086
7-Methoxy-1-tetralone 97%
7-Methoxy-1-tetralone
Formula: C₁₁H₁₂O₂
Melting Pt: 59…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00001696
CAS Number: 6836-19-7
EINECS: 229-916-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR