4-Bromo-N,N-diphenylaniline 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR17665-100G OR17665-25G
77406-074EA 110.04 USD
77406-074 77406-076
4-Bromo-N,N-diphenylaniline 98%
4-Bromo-N,N-diphenylaniline
Formula: C₁₈H₁₄BrN
Melting Pt: 108…112 °C
Storage Temperature: Ambient
MDL Number: MFCD01851266
CAS Number: 36809-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR