1H-Pyrrolo[2,3-d]pyrimidin-4(7H)-one 95+%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 7-Deazahypoxanthine

OR17055-1G OR17055-25G OR17055-5G
77405-926EA 21.26 USD
77405-926 77405-928 77405-930
1H-Pyrrolo[2,3-d]pyrimidin-4(7H)-one 95+%
1H-Pyrrolo[2,3-d]pyrimidin-4(7H)-one
Formula: C₆H₅N₃O
MW: 135.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08234923
CAS Number: 3680-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR