4-(3-Pyridyl)aniline

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Pyridin-3-ylaniline

OR2020-1G OR2020-250MG
77406-558EA 371.86 USD
77406-558 77406-560
4-(3-Pyridyl)aniline
4-(3-Pyridyl)aniline
Formula: C₁₁H₁₀N₂
MW: 170.22 g/mol
Melting Pt: 115…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00956763
CAS Number: 82261-42-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR