2-(Methylsulfonyl)aniline 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Aminophenyl methyl sulphone

OR16499-1G OR16499-25G OR16499-5G
77405-720EA 38.72 USD
77405-720 77405-722 77405-724
2-(Methylsulfonyl)aniline 99%
2-(Methylsulfonyl)aniline
Formula: C₇H₉NO₂S
MW: 171.217 g/mol
Melting Pt: 84…85 °C
Storage Temperature: Ambient
MDL Number: MFCD08444390
CAS Number: 2987-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR