1,1'-Bis(di-tert-butylphosphino)ferrocene 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1,1'-Bis(di-t-butylphosphino)ferrocene, 1,1'-Bis[bis(1,1-dimethylethyl)phosphino]ferrocene, DTBPF

OR16471-1G OR16471-5G OR16471-10G
77405-714EA 21.26 USD
77405-714 77405-716 77405-712
1,1'-Bis(di-tert-butylphosphino)ferrocene 98%
1,1'-Bis(di-tert-butylphosphino)ferrocene
Formula: C₂₆H₄₄FeP₂
MW: 474.42 g/mol
Melting Pt: 73…75 °C
Storage Temperature: Ambient
MDL Number: MFCD01630818
CAS Number: 84680-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR