2-Hydroxy-6-methylbenzoic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,6-Cresotic acid, 6-Hydroxy-o-toluic acid, 6-Methylsalicylic acid

OR16150-10G OR16150-1G OR16150-5G
77405-646EA 95.76 USD
77405-646 77405-648 77405-650
2-Hydroxy-6-methylbenzoic acid 95%
2-Hydroxy-6-methylbenzoic acid
Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 170…171 °C
Storage Temperature: Ambient
MDL Number: MFCD01194284
CAS Number: 567-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR