Oxo[4-(piperidin-1-yl)phenyl]acetaldehyde hydrate 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-[4-(2-Oxoethanoyl)phenyl]piperidine hydrate, 4-(Piperidin-1-yl)phenylglyoxal hydrate

OR1613-1G OR1613-500MG OR1613-5G
77405-634EA 166.06 USD
77405-634 77405-636 77405-638
Oxo[4-(piperidin-1-yl)phenyl]acetaldehyde hydrate 95%
Oxo[4-(piperidin-1-yl)phenyl]acetaldehyde hydrate
Formula: C₁₃H₁₅NO₂.H₂O
Storage Temperature: Ambient
MDL Number: MFCD08705865
CAS Number: 93290-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR