3-Amino-5-azaindole dihydrochloride

Supplier: APOLLO SCIENTIFIC
Warning

OR15771-1G OR15771-500MG
77405-568EA 929.16 USD
77405-568 77405-570
3-Amino-5-azaindole dihydrochloride
3-Amino-5-azaindole dihydrochloride
Formula: C₇H₇N₃·2ClH
Storage Temperature: Refrigerator
MDL Number: MFCD16628055
CAS Number: 1257535-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR