5,6-Dichloro-1H-indole

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5,6-Dichloroindole

OR15747-100G OR15747-25G OR15747-5G
77405-556EA 908.78 USD
77405-556 77405-558 77405-560
5,6-Dichloro-1H-indole
5,6-Dichloro-1H-indole
Formula: C₈H₅Cl₂N
MW: 186.0400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01075036
CAS Number: 121859-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR