N-(tert-Butoxycarbonyl)-L-propargylglycine 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)pent-4-ynoic acid, N-Boc-L-propargylglycine, (S)-N-BOC-Propargylglycine, Boc-propargyl-Gly-OH, (S)-2-(Boc-amino)-4-pentynoic Acid, Boc-Pra-OH, (S)-2-(tert-Butoxycarbonylamino)-4-pentynoic Acid

OR14674-10G OR14674-25G OR14674-5G
77405-046EA 273.04 USD
77405-046 77405-048 77405-050
N-(tert-Butoxycarbonyl)-L-propargylglycine 97%
N-(tert-Butoxycarbonyl)-L-propargylglycine
Formula: C₁₀H₁₅NO₄
MW: 213.23 g/mol
Melting Pt: 85 °C
Storage Temperature: Freezer
MDL Number: MFCD01320855
CAS Number: 63039-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR