Methyl 3,5-dinitro-2-hydroxybenzoate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,4-Dinitro-6-(methoxycarbonyl)phenol, Methyl 3,5-dinitrosalicylate

OR14438-1G OR14438-5G
77404-966EA 38.72 USD
77404-966 77404-968
Methyl 3,5-dinitro-2-hydroxybenzoate
Methyl 3,5-dinitro-2-hydroxybenzoate
Formula: C₈H₆N₂O₇
MW: 242.142 g/mol
Melting Pt: 125…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00017561
CAS Number: 22633-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR