1,3-Benzothiazole-6-sulfonyl chloride 95%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
Benzo[d]thiazole-6-sulphonyl chloride, 6-(Chlorosulphonyl)-1,3-benzothiazole
OR14303-1G
OR14303-250MG
77404-890EA
55.02
USD
77404-890
77404-892
1,3-Benzothiazole-6-sulfonyl chloride 95%
1,3-Benzothiazole-6-sulfonyl chloride
Formula:
C₇H₄ClNO₂S₂ MW: 233.695 g/mol Melting Pt: 118…119 °C Storage Temperature: Ambient |
MDL Number:
MFCD03086100 CAS Number: 181124-40-3 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...