6-Methylnicotinic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 6-Methyl-3-pyridinecarboxylic acid, 6-Methylpyridine-3-carboxylic acid

OR13706-100G OR13706-1G OR13706-25G
77404-710EA 124.3 USD
77404-710 77404-712 77404-714
6-Methylnicotinic acid 98%
6-Methylnicotinic acid
Formula: C₇H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD00006341
CAS Number: 3222-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR