Bzl-Pro-OH
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
(2S)-1-Benzylpyrrolidine-2-carboxylic acid, (S)-1-Benzylpyrrolidine-2-carboxylic acid
OR13357-1G
OR13357-5G
77404-630EA
55.02
USD
77404-630
77404-632
Bzl-Pro-OH
Bzl-Pro-OH
Formula:
C₁₂H₁₅NO₂ MW: 205.3 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00235894 CAS Number: 31795-93-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...