4-(Acetamidomethyl)benzeneboronic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-[(Acetylamino)methyl]benzeneboronic acid, (4-(Acetamidomethyl)phenyl)boronic acid

OR1333-100MG OR1333-250MG
77404-624EA 24.36 USD
77404-624 77404-626
4-(Acetamidomethyl)benzeneboronic acid 97%
4-(Acetamidomethyl)benzeneboronic acid
Formula: C₉H₁₂BNO₃
MW: 193.01 g/mol
Melting Pt: 278…280 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06659818
CAS Number: 850568-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR