2-Chloroethyl phenyl sulfone 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: [(2-Chloroethyl)sulphonyl]benzene

OR1291-100G OR1291-25G OR1291-5G
77404-468EA 666.3 USD
77404-468 77404-470 77404-472
2-Chloroethyl phenyl sulfone 98%
2-Chloroethyl phenyl sulfone
Formula: C₈H₉ClO₂S
Boiling Pt: 168…174 °C/7 mmHg
Melting Pt: 52…55 °C
Storage Temperature: Ambient
MDL Number: MFCD00025047
CAS Number: 938-09-0
EINECS: 213-337-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR