2,5-Dichloropyrazine

Supplier: APOLLO SCIENTIFIC
Warning

OR16998-250MG OR16998-1G
77405-894EA 21.32 USD
77405-894 77405-892
2,5-Dichloropyrazine
2,5-Dichloropyrazine
Formula: C₄H₂Cl₂N₂
MW: 148.9781 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04117886
CAS Number: 19745-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR