4-(4-Nitrophenyl)piperidine 98+%

Supplier: APOLLO SCIENTIFIC
Warning

OR15358-100MG OR15358-250MG
77405-378EA 21.26 USD
77405-378 77405-380
4-(4-Nitrophenyl)piperidine 98+%
4-(4-Nitrophenyl)piperidine
Formula: C₁₁H₁₄N₂O₂
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD06804549
CAS Number: 26905-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR