2-Bromo-4,6-dimethylbenzenol 96%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Bromo-4,6-dimethylphenol

OR15140-1G OR15140-25G OR15140-5G
77405-310EA 41.78 USD
77405-310 77405-312 77405-314
2-Bromo-4,6-dimethylbenzenol 96%
2-Bromo-4,6-dimethylbenzenol
Formula: C₈H₉BrO
MW: 201.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08443978
CAS Number: 15191-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR