4-(Isopropylamino)diphenylamine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: N1-Isopropyl-N4-phenylbenzene-1,4-diamine, N-Isopropyl-N'-phenyl-p-phenylenediamine

OR15007-100G OR15007-25G
77405-230EA 28.4 USD
77405-230 77405-232
4-(Isopropylamino)diphenylamine
4-(Isopropylamino)diphenylamine
Formula: C₁₅H₁₈N₂
Boiling Pt: 161 °C/1 mmHg
Melting Pt: 74…80 °C
Density: 1.04 g/cm³ (25 °C)
Flash Pt: 172 °C
Storage Temperature: Ambient
MDL Number: MFCD00026348
CAS Number: 101-72-4
EINECS: 202-969-7
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR