4-Bromo-m-anisidine 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
4-Bromo-3-methoxyaniline, 5-Amino-2-bromoanisole, 4-Bromo-3-anisidine
OR15001-100G
OR15001-25G
OR15001-5G
77405-218EA
103.02
USD
77405-218
77405-220
77405-222
4-Bromo-m-anisidine 97%
4-Bromo-m-anisidine
Formula:
C₇H₈BrNO Melting Pt: 96…100 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD05664063 CAS Number: 19056-40-7 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...