3-Bromo-4-(hydroxymethyl)thiophene 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR12445-1G OR12445-250MG
77404-308EA 234.32 USD
77404-308 77404-310
3-Bromo-4-(hydroxymethyl)thiophene 97%
3-Bromo-4-(hydroxymethyl)thiophene
Formula: C₅H₅BrOS
Melting Pt: 53…53.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01859831
CAS Number: 70260-05-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR