5-Methylthien-2-yl isocyanate 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR14873-1G OR14873-250MG OR14873-500MG
77405-124EA 192.56 USD
77405-124 77405-126 77405-128
5-Methylthien-2-yl isocyanate 97%
5-Methylthien-2-yl isocyanate

May form small amount of sediment.

Formula: C₆H₅NOS
Boiling Pt: 94 °C (53 mmHg)
Storage Temperature: Refrigerator
MDL Number: MFCD09817547
CAS Number: 76536-99-7
UN: 2206
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR